Cole-Parmer® Essentials Stainless Steel Graduated Pouring Beakers
Easily measure and pour samples
  • Measure volumes easily with internal graduation markers
  • Great for higher temperatures— withstand temperatures up to 650°F (182° C)
These graduated pouring beakers Features easy grip handles for convenient pouring. Beakers are graduated in mL and oz and constructed of medium gauge 304 stainless steel.
£104.58 - £230.61GBP / Each
£125.50 - £276.73GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
500 Stainless Steel 3 3/4
£104.58GBP / Each
£125.50GBP (incl VAT)
COMPARE
1000 Stainless Steel 4 3/4
£165.31GBP / Each
£198.37GBP (incl VAT)
COMPARE
2000 Stainless Steel 5 1/2
£205.88GBP / Each
£247.06GBP (incl VAT)
COMPARE
5000 Stainless Steel 7
£230.61GBP / Each
£276.73GBP (incl VAT)