Cole-Parmer® Solvent-Resistant PVC 2-Stop Pump Tubing
Phthalate-free PVC with exceptional chemical resistance to oils and aromatic solvents
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Excellent pumping life for routine applications
  • Exhibits very low gas permeability
  • Translucent tubing allows for visual confirmation of fluid flow
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use solvent-resistant PVC tubing with aromatic oils, solvents and petroleum products. It exhibits similar chemical resistance characteristics to Viton® tubing, but at a lower cost, making it an excellent alternative. The phthalate-free and anti-leaching characteristics minimize the risk of interferences or false positives, ensuring accurate and reliable results. Translucent tubing allows for easy visual confirmation of fluid flow. Bright yellow tubing helps differentiate from standard PVC stop tubing. Solvent-resistant PVC tubing has a temperature ranging from -50 to 74°C (-58 to 165°F).
£43.12 - £45.49GBP / Pkg of 12
£51.74 - £54.59GBP (incl VAT)
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Red 0.19
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Orange/Blue 0.25
£43.12GBP / Pkg of 12
£51.74GBP (incl VAT)
COMPARE
Orange/Green 0.38
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Orange/Yellow 0.51
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Orange/White 0.64
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Black/Black 0.76
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Orange/Orange 0.89
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
White/White 1.02
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Red/Red 1.14
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Gray/Gray 1.3
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Yellow/Blue 1.52
£45.49GBP / Pkg of 12
£54.59GBP (incl VAT)
COMPARE
Blue/Blue 1.65
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Green/Green 1.85
£44.45GBP / Pkg of 12
£53.34GBP (incl VAT)
COMPARE
Purple/Purple 2.06
£43.12GBP / Pkg of 12
£51.74GBP (incl VAT)
COMPARE
Purple/Black 2.29
£43.12GBP / Pkg of 12
£51.74GBP (incl VAT)
COMPARE
Purple/Orange 2.54
£45.49GBP / Pkg of 12
£54.59GBP (incl VAT)
COMPARE
Purple/White 2.79
£43.12GBP / Pkg of 12
£51.74GBP (incl VAT)
Showing 1- 17 of 17