Cole-Parmer® Sanitary Gaskets, Platinum-Cured Silicone
Compatible with Tri-Clamp® style fittings
  • Great for most pharmaceutical, food, and beverage applications
  • Usable temperature range: –40 to 232 °C (–40 to 450 °F)
  • Animal-derived ingredient free and phthalate free
  • FDA CFR 21 177 2600, USP Class VI, and 3A compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Use the platinum-cured silicone gaskets in sanitary fittings for leak-free fluid transfer. These nontoxic and highly biocompatible gaskets meet FDA, USP, and 3A compliance, making them ideal for pharmaceutical, food, and beverage applications. Gaskets are free of phthalates, TSE/BSE, and animal-derived ingredients, ensuring the integrity of your fluid.
£22.85 - £30.05GBP / Pkg of 10
£27.42 - £36.06GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sanitary Gasket Size (in) Seal Material Description
Availability Pricing
COMPARE
1/2 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 1/2"; 10/Pk
£28.80GBP / Pkg of 10
£34.56GBP (incl VAT)
COMPARE
3/4 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 3/4"; 10/Pk
£26.94GBP / Pkg of 10
£32.33GBP (incl VAT)
COMPARE
1 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 1"; 10/Pk
£30.05GBP / Pkg of 10
£36.06GBP (incl VAT)
COMPARE
1 1/2 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 1-1/2"; 10/Pk
£22.85GBP / Pkg of 10
£27.42GBP (incl VAT)
COMPARE
2 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 2"; 10/Pk
£24.09GBP / Pkg of 10
£28.91GBP (incl VAT)
COMPARE
2 1/2 Platinum-Cured Silicone Sanitary Gasket, Platinum-Cured Silicone, 2-1/2"; 10/Pk
£28.80GBP / Pkg of 10
£34.56GBP (incl VAT)