Cole-Parmer® Rectangular Carboy with Spigot, PP
Maintain sample integrity
  • Leakproof safe transport
  • USP Class VI certified
  • Autoclavable
These leakproof polypropylene (PP) rectangular carboys with spigot easily store and dispense sterile water and other solutions. Space-saving design requires least space than traditional round carboys. Featrues include built-in stainless steel handle and wide mouth opening for easy cleaning.
£201.60 - £217.58GBP / Each
£241.92 - £261.10GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Overall Height (in)
Availability Pricing
COMPARE
20 Polypropylene(PP) 19
£217.58GBP / Each
£261.10GBP (incl VAT)
COMPARE
9 Polypropylene(PP) 15 19/64
£201.60GBP / Each
£241.92GBP (incl VAT)