Cole-Parmer® PVC 3-Stop Pump Tubing with Flared Ends
Flared end allows easy connection between the sample and peristaltic tubing
  • Stops ensure a perfect fit every time
  • Chemically resistant to a wide range of solutions
  • Delivers accurate and interference-free analytical results
  • Color-coded stops for easy identification
  • Transparent—easily view fluid flow
  • Certified to USP Class VI, USP37-NF32 2014
  • Meets European Pharmacopoeia Monograph 3.1.2
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Flared tubing provides the same durability and performance as regular PVC tubing, but with the added convenience of flared ends to make installation easier, especially for small-bore tubing. Made from phthalate-free PVC, the tubing exhibits great resistance to a wide range of common chemicals, ensuring dependable performance for many routine applications. Anti-leaching characteristics minimize the risk of interferences or false positives, ensuring accurate and reliable results. Easily view fluid flow through the transparent PVC for easy observation of pump performance. PVC tubing withstands temperatures ranging from -50 to 74°C (-58 to 165°F).
£71.86 - £77.18GBP / Pkg of 12
£86.23 - £92.62GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Red 0.19
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Orange/Blue 0.25
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Orange/Green 0.38
£77.18GBP / Pkg of 12
£92.62GBP (incl VAT)
COMPARE
Green/Yellow 0.44
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Orange/Yellow 0.51
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Orange/White 0.64
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Black/Black 0.76
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Orange/Orange 0.89
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
White/White 1.02
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Red/Red 1.14
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Gray/Gray 1.3
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Yellow/Yellow 1.42
£72.67GBP / Pkg of 12
£87.20GBP (incl VAT)
COMPARE
Yellow/Blue 1.52
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Blue/Blue 1.65
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Green/Green 1.85
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Purple/Purple 2.06
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Purple/Black 2.29
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Purple/Orange 2.54
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Purple/White 2.79
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
COMPARE
Black/White 3.18
£71.86GBP / Pkg of 12
£86.23GBP (incl VAT)
Showing 1- 20 of 20