Cole-Parmer® Powder Funnels, PP and HDPE
Prevent blockages with wide base opening
  • Constructed from durable food grade plastic
  • Cleanroom class 100,000 packed and individually bagged
  • Disposable funnels are ideal for food and pharmaceutical applications
These Cole-Parmer® essentials powder funnels are ideal for transferring powders and granules. Disposable funnels are packaged in class 100,000 cleanroom and individually bagged to avoid cross contamination. The wide opening at the base of the funnel will help prevent powder blockages.
£108.69 - £283.74GBP / Pkg of 10
£130.43 - £340.49GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Top OD (mm) Description
Availability Pricing
COMPARE
HDPE 200 Powder Funnel, HDPE, 200 mm Diameter; 10/Pk
£108.69GBP / Pkg of 10
£130.43GBP (incl VAT)
COMPARE
HDPE 200 Powder Funnel, HDPE, Sterile, 200 mm Diameter; 10/Pk
£181.28GBP / Pkg of 10
£217.54GBP (incl VAT)
COMPARE
Polypropylene 350 Powder Funnel, Polypropylene, 350 mm Diameter; 8/Pk
£182.11GBP / Pkg of 8
£218.53GBP (incl VAT)
COMPARE
Polypropylene 350 Powder Funnel, Polypropylene, Sterile, 350 mm Diameter; 8/Pk
£283.74GBP / Pkg of 8
£340.49GBP (incl VAT)