Cole-Parmer® Polycarbonate Storage Boxes
Designed to withstand temperatures from -320.8 to 250°F (-196 to 121°C)
  • Clear lids with locator grids simplify sample identification
  • Keyed lid prevents misalignment of grid
Polycarbonate freezer boxes, available with 25, 81, and 100-place divisions, feature bottom drain holes for safe venting. The 25- and 81-place boxes have individual cell compartment dividers, while the 100-place box holds tubes in a pegged grid divider.
£66.17 - £110.82GBP / Pkg of 4
£79.40 - £132.98GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Freezer Box, PC, 100-Place, 2", 10.5 mm; 4/Pk
£87.39GBP / Pkg of 4
£104.87GBP (incl VAT)
COMPARE
Freezer Box, PC, 81-Place, 2", 12.5 mm; 4/Pk
£89.02GBP / Pkg of 4
£106.82GBP (incl VAT)
COMPARE
Freezer Box, PC, 81-Place, 3", 12.5 mm; 4/Pk
£110.82GBP / Pkg of 4
£132.98GBP (incl VAT)
COMPARE
Freezer Box, PC, 25-Place, 2", 12.5 mm; 4/Pk
£66.17GBP / Pkg of 4
£79.40GBP (incl VAT)