Cole-Parmer® Peroxide-Cured Silicone 3-Stop Pump Tubing
Inhibits microbiological growth—excellent for pharmaceutical and biological research applications
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Exhibits good heat, cold, chemical, and abrasion resistance
  • USP Class VI, REACH, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
The high-performance, peroxide-cured silicone tubing features a smooth bore to inhibit microbial growth and effortless fluid transfer. Enhanced mechanical strength and high elasticity make the tubing ideal for use in peristaltic pumps. Tubing also exhibits a wide operating temperature range, excellent chemical resistance, and kink resistance, making it great for various applications. Translucent tubing offers easy visual confirmation of fluid flow. Silicone tubing has a wide operating temperature range from -50 to 230°C (-58 to 446°F).
£48.45 - £54.30GBP / Pkg of 12
£58.14 - £65.16GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Orange 0.89
£48.45GBP / Pkg of 12
£58.14GBP (incl VAT)
COMPARE
Red/Red 1.14
£54.30GBP / Pkg of 12
£65.16GBP (incl VAT)