Cole-Parmer® Ovine and Caprine Infection Detection Kits for qPCR
Quickly and accurately detect the presence of ovine and caprine infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Capripoxvirus No DNA
Call For Price
COMPARE
Peste des Petits Ruminants Virus No RNA
Call For Price
COMPARE
Peste des Petits Ruminants Virus Yes RNA
Call For Price
COMPARE
Sheep Poxvirus No DNA
Call For Price
COMPARE
Sheep Poxvirus Yes DNA