Cole-Parmer® Omega Zen™ Pipette Controller Accessories
Enhance and maintain your Omega Zen pipettors with these accessories
  • Compatible with Omega Zen pipette controllers
  • Choose from a wide range of accessories to make your pipettor even more versatile
These accessories can be used to enhance the flexibility of your Omega Zen controller, or be used for routine maintenance. There are four filter options to choose from. All are made of a polypropylene housing with a hydrophobic PTFE membrane. The Omega Zen controllers accept either the 0.2 µm or the 0.45 µm pore size filters.
Power your Omega Zen controller with the rechargeable lithium-ion battery for up to 9 hours of continuous use and charge the battery with the universal power supply.
The nosepiece and silicone pipette adapter are both autoclavable for optional sterility.
£16.38 - £79.27GBP / Pkg of 10
£19.66 - £95.12GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Body Material Description
Availability Pricing
COMPARE
Plastic Pipettor Battery Charging Desk Stand
£54.52GBP / Each
£65.42GBP (incl VAT)
COMPARE
Plastic Pipettor Standard Wall-Mount
£16.38GBP / Each
£19.66GBP (incl VAT)
COMPARE
Plastic Pipettor Margnetic Wall-Mount
£21.84GBP / Each
£26.21GBP (incl VAT)
COMPARE
Hydrophobic PTFE Filters, 0.2 µm, Sterile; 10/pk
£54.87GBP / Pkg of 10
£65.84GBP (incl VAT)
COMPARE
Hydrophobic PTFE Filters, 0.2 µm, Non-sterile; 10/pk
£41.07GBP / Pkg of 10
£49.28GBP (incl VAT)
COMPARE
Hydrophobic PTFE Filters, 0.45 µm, Sterile; 10/pk
£79.27GBP / Pkg of 10
£95.12GBP (incl VAT)
COMPARE
Hydrophobic PTFE Replacement Filters, 0.45 µm, Non-sterile; 10/pk
£53.39GBP / Pkg of 10
£64.07GBP (incl VAT)
COMPARE
Replacement Power Supply
£39.36GBP / Each
£47.23GBP (incl VAT)
COMPARE
Replacement Battery; Lithium-Ion
£36.26GBP / Each
£43.51GBP (incl VAT)
COMPARE
UV-resistant plastic Replacement Nosepiece
£27.55GBP / Each
£33.06GBP (incl VAT)
COMPARE
Silicone Replacement Silicone Pipette Adapter
£16.38GBP / Each
£19.66GBP (incl VAT)
Showing 1- 11 of 11