Cole-Parmer® Miniature Check Valves, Hose Barb, Polypropylene and Viton®
Versatile check valves can be installed in any position
  • Prevent backflow to protect your equipment
  • Work with both liquids and gases
  • Use for pressure or vacuum applications
  • 0.18 psi breaking pressure
Install these miniature check valves with hose barbs in any position to prevent backflow. They are compatible with both liquids and gases for pressure or vacuum applications. They require 0.18 psi breaking pressure; closing pressure is 0.014 psi.
£3.11 - £4.10GBP / Each
£3.73 - £4.92GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Coefficient (Cv) Inlet Port Outlet Port
Availability Pricing
COMPARE
0.11 1/8" hose barb 1/8" hose barb
£3.11GBP / Each
£3.73GBP (incl VAT)
COMPARE
0.12 3/16" hose barb 3/16" hose barb
£3.71GBP / Each
£4.45GBP (incl VAT)
COMPARE
0.13 5/16" hose barb 5/16" hose barb
£4.10GBP / Each
£4.92GBP (incl VAT)
COMPARE
0.12 1/4" hose barb 1/4" hose barb
£4.10GBP / Each
£4.92GBP (incl VAT)