Cole-Parmer® Long-Handled Lab Sampling Scoops, HDPE
Extra-long handle makes reaching into deep containers easier, keeps hands further away from sample
  • Manufactured in an ISO 8 (Class 100,000) cleanroom to ensure cleanliness and reduce cross-contamination
  • High-quality HDPE conforms to FDA, EC, and EU regulations
  • BSE/TSE free
Use these long-handled, non-sterile, disposable lab scoops to gather samples such as powders and granules. The 300 cm (11.8 in) long handle allows you to reach into deep containers, while keeping your hands away from the materials being measured. The long handle also makes this lab scoop more visible when left in a container. Manufactured of high-density polyethylene (HDPE) in an ISO 8 (Class 100,000) clean room, these sampling scoops withstand exposure to most chemicals without corroding. They are individually packaged in a heat-sealed PE/PET bag and can be used straight out of the bag without cleaning.
These laboratory scoops conform to FDA CFR 177.1520, EU 10/2011, and EC 1935/2004 regulations, and are BSE/TSE free. Indicated capacities are approximate and are not intended for measuring specific volumes.
£159.92 - £165.10GBP / Pkg of 50
£191.90 - £198.12GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
50 37.1 4
£159.92GBP / Pkg of 50
£191.90GBP (incl VAT)
COMPARE
100 39 5
£165.10GBP / Pkg of 50
£198.12GBP (incl VAT)