Cole-Parmer® Liquid Nitrogen Dewars
Safely store and dispense liquid nitrogen
  • Ribbed aluminum body and magneformed necktube offer durability and structural strength
  • Superior insulation ensures high thermal efficiency
  • Lightweight and handles for easy handling
Use these liquid nitrogen (LN2) dewars to safely store and dispense small quantities of LN2. Rugged, ribbed aluminum body and magneformed neck tube provide excellent durability for use in lab or industrial environments. High thermal efficiency insulation minimizes nitrogen loss and provides long holding times. Handles on all models make it easy to handle. Durable paint finish resists wear and corrosion.
£945.78 - £1,792.00GBP / Each
£1,134.94 - £2,150.40GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Evaporation (Liters/day) Static Holding Time (days) Volume (Liters)
Availability Pricing
COMPARE
0.22 45 10
£945.78GBP / Each
£1,134.94GBP (incl VAT)
COMPARE
0.23 109 25
£1,265.19GBP / Each
£1,518.23GBP (incl VAT)
COMPARE
0.29 120 35
£1,410.38GBP / Each
£1,692.46GBP (incl VAT)
COMPARE
0.49 102 50
£1,792.00GBP / Each
£2,150.40GBP (incl VAT)