Cole-Parmer® Ladles, Sterile
Reduce cross contamination while sampling liquids
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Designed to go through traditional 2” bungs
These ladles are designed to easily take liquid samples with the added convenience of not having to clean the sampler prior to use. The ladles are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Samplers are sterilized via gamma irradiation (25 to 45 kGy). Choose from polypropylene or high-density polyethylene (HDPE) construction for a wide range of applications.
£160.75GBP / Pkg of 40
£192.90GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Material Volume (mL)
Availability Pricing
COMPARE
45.3 Polypropylene 100
£160.75GBP / Pkg of 40
£192.90GBP (incl VAT)
COMPARE
26 HDPE 50
£160.75GBP / Pkg of 40
£192.90GBP (incl VAT)
COMPARE
26 Polypropylene 50
£160.75GBP / Pkg of 40
£192.90GBP (incl VAT)