Cole-Parmer® Ladder Cable Zip Ties, White Nylon
Flexible ties bend easily
  • Conforms to bundle shape
  • Locks strongly for a secure hold
  • Withstands temperatures to 85 °C (185 °F)
Made of high-quality 6/6 nylon, these ladder style cable zip ties are strong, yet flexible enough to tightly conform to irregular shaped bundles. The durable locking mechanism holds securely, even when used to bundle vibrating or flexing materials. They will not corrode or stretch and will continue to hold tension even in oily spray or wet environments. A bent tail allows for easy pickup of the tie and threading through the lock.
Able to withstand temperatures to 85 °C (185 °F), these cable/zip ties have a melting point of 257 °C (495 °F) and remain flexible to -40 °C (-40 °F). Efficient range of ties may be higher or lower in your specific application. They are RoHS compliant, UL recognized, and UL 94 V-2 flammability rated.
Easily install by wrapping the tie around the bundle and threading the tail through the locking mechanism. Pull tight by hand and cut the tail to complete or use a tensioning tool—available separately. For mounting wires and cables to walls or other surfaces, use Cole-Parmer® Self Adhering Cable Zip Tie Mounts —available separately.
£55.67 - £74.17GBP / Bag of 1000
£66.80 - £89.00GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Bundle Diameter (in) Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
2 1/10 7 1/2 40
£55.67GBP / Bag of 1000
£66.80GBP (incl VAT)
COMPARE
3 11 1/2 40
£74.17GBP / Bag of 1000
£89.00GBP (incl VAT)