Cole-Parmer® Instrument Tees with 1-1/2” Sanitary Connection
Built-in elastomer diaphragm makes single-use pressure sensing simple
  • Accepts pressure sensor with standard 1-1/2" sanitary connection
  • Fused diaphragm creates a sterile barrier
  • No solvents or adhesives used for the diaphragm
  • Excellent biocompatibility and low extractables
  • ASME-BPE, USP Class VI, REACH, and RoHS compliant
  • Sterilize by autoclave, gamma irradiation, or EtO
Easily measure the pressure in your line while maintaining sterility with the instrument tees. Each tee features a sanitary connection with a fused elastomer diaphragm without solvents or adhesives, resulting in a single-piece unit, helping to maintain barrier integrity throughout operations. The 1-1/2” sanitary connection provides superior ergonomics and exact alignment, ensuring accurate pressure measurements.
To ensure quality and integrity, the instruments tees are manufactured in controlled environment and packaged in ISO Class 7 cleanroom.
£161.30 - £255.19GBP / Each
£193.56 - £306.23GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port
Availability Pricing
COMPARE
1/2" mono barb 1/2" mono barb
£161.30GBP / Each
£193.56GBP (incl VAT)
COMPARE
1/2" mono barb 1/2" mini tri clamp
£207.04GBP / Each
£248.45GBP (incl VAT)
COMPARE
1/2" mini tri clamp 1/2" mini tri clamp
£252.23GBP / Each
£302.68GBP (incl VAT)
COMPARE
1" mono barb 1" tri clamp
£255.19GBP / Each
£306.23GBP (incl VAT)
COMPARE
1/2" mono barb 1/2" mono barb
£161.30GBP / Each
£193.56GBP (incl VAT)
COMPARE
1/2" mono barb 1/2" mini tri clamp
£207.04GBP / Each
£248.45GBP (incl VAT)
COMPARE
1/2" mini tri clamp 1/2" mini tri clamp
£252.23GBP / Each
£302.68GBP (incl VAT)
COMPARE
1" mono barb 1" mono barb
£209.45GBP / Each
£251.34GBP (incl VAT)
Showing 1- 8 of 8