Cole-Parmer® Hose Clamps, Acetal
Fast, secure, and reusable hose clamps
  • Installs up to five times faster than conventional clamps
  • One-hand installation is perfect for tight spaces
  • Heat-stabilized for high-temperature applications (up to 200°F / 93°C)
  • Resistant to rust, corrosion, and a wide range of chemicals
Cole-Parmer® Acetal Hose Clamps revolutionize hose connections with ratchet-style closure, allowing for quick and secure fastening. Its reusable, nonconductive white acetal copolymer ensures long-lasting durability without the risk of rust or corrosion. Engineered for safety and efficiency, it features a smooth, low-profile design with no protruding bands. Designed for under-hood and industrial applications, this clamp stays put once set, handling everything from lubricants and gasoline to detergents and salts, bases, alcohols or ethers.
£37.04 - £78.34GBP / Pkg of 100
£44.45 - £94.01GBP (incl VAT)
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Tubing Size Range
Availability Pricing
COMPARE
0.28 - 0.30"
£44.52GBP / Pkg of 100
£53.42GBP (incl VAT)
COMPARE
0.31 - 0.35"
£40.38GBP / Pkg of 100
£48.46GBP (incl VAT)
COMPARE
0.57 - 0.65"
£64.97GBP / Pkg of 100
£77.96GBP (incl VAT)
COMPARE
0.24" - 0.26"
£40.38GBP / Pkg of 100
£48.46GBP (incl VAT)
COMPARE
0.61 - 0.72"
£44.52GBP / Pkg of 100
£53.42GBP (incl VAT)
COMPARE
0.71 - 0.81"
£45.26GBP / Pkg of 100
£54.31GBP (incl VAT)
COMPARE
0.78 - 0.93"
£45.26GBP / Pkg of 100
£54.31GBP (incl VAT)
COMPARE
0.88 - 1.0"
£53.49GBP / Pkg of 100
£64.19GBP (incl VAT)
COMPARE
0.35 - 0.40"
£38.75GBP / Pkg of 100
£46.50GBP (incl VAT)
COMPARE
0.92 - 1.05"
£53.49GBP / Pkg of 100
£64.19GBP (incl VAT)
COMPARE
1.0 - 1.12"
£53.49GBP / Pkg of 100
£64.19GBP (incl VAT)
COMPARE
1.11 - 1.27"
£54.30GBP / Pkg of 100
£65.16GBP (incl VAT)
COMPARE
0.39 - 0.44"
£44.52GBP / Pkg of 100
£53.42GBP (incl VAT)
COMPARE
1.21 - 1.38"
£78.34GBP / Pkg of 100
£94.01GBP (incl VAT)
COMPARE
0.38 - 0.42"
£37.04GBP / Pkg of 100
£44.45GBP (incl VAT)
COMPARE
0.45 - 0.51"
£37.86GBP / Pkg of 100
£45.43GBP (incl VAT)
Showing 1- 16 of 16