Cole-Parmer® Hose Barb Fittings, Straight Reducer, Natural Polycarbonate
Easily connect two different size tubing
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Create a smooth, in-line transition between different tubing sizes using straight reducers. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
£5.04 - £5.36GBP / Pkg of 10
£6.05 - £6.43GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in)
Availability Pricing
COMPARE
3/32 1/16
£5.36GBP / Pkg of 10
£6.43GBP (incl VAT)
COMPARE
1/8 1/16
£5.36GBP / Pkg of 10
£6.43GBP (incl VAT)
COMPARE
1/8 3/32
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
5/32 1/16
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
5/32 1/8
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
5/32 3/32
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
3/16 1/16
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
3/16 1/8
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
3/16 3/32
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
3/16 5/32
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
1/4 1/8
£5.04GBP / Pkg of 10
£6.05GBP (incl VAT)
COMPARE
1/4 3/16
£5.19GBP / Pkg of 10
£6.23GBP (incl VAT)
COMPARE
1/4 5/32
£5.19GBP / Pkg of 10
£6.23GBP (incl VAT)
Showing 1- 13 of 13