Cole-Parmer® Hose Barb Fitting, Straight Reducer, HDPE
Straight reducer hose barb fitting delivers a secure, leak-resistant connection between different hose sizes.
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520 (when used unmodified for the manufacture of food contact articles) and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Easily connect two different size tubing with straight reducers. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
£5.70 - £12.90GBP / Pkg of 10
£6.84 - £15.48GBP (incl VAT)
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/32 1/16 HDPE
£5.70GBP / Pkg of 10
£6.84GBP (incl VAT)
COMPARE
1/8 1/16 HDPE
£5.70GBP / Pkg of 10
£6.84GBP (incl VAT)
COMPARE
1/8 3/32 HDPE
£5.70GBP / Pkg of 10
£6.84GBP (incl VAT)
COMPARE
5/32 3/32 HDPE
£5.70GBP / Pkg of 10
£6.84GBP (incl VAT)
COMPARE
5/32 1/8 HDPE
£7.00GBP / Pkg of 10
£8.40GBP (incl VAT)
COMPARE
3/16 3/32 HDPE
£5.70GBP / Pkg of 10
£6.84GBP (incl VAT)
COMPARE
3/16 1/8 HDPE
£7.00GBP / Pkg of 10
£8.40GBP (incl VAT)
COMPARE
1/4 3/32 HDPE
£10.05GBP / Pkg of 10
£12.06GBP (incl VAT)
COMPARE
1/4 5/32 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/4 3/16 HDPE
Call For Price
COMPARE
5/16 5/32 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
5/16 1/4 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 1/8 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 5/32 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 1/4 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
3/8 5/16 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/2 1/4 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/2 5/16 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
1/2 3/8 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
COMPARE
5/8 1/4 HDPE
£9.65GBP / Pkg of 10
£11.58GBP (incl VAT)
Showing 1 - 20 of 29