Cole-Parmer® Heavy-Walled Narrow Mouth Carboys, HDPE
Heavy wall for added durability—ship without worry!
  • Chemical resistant
  • Handle allows for easy carrying of solutions
Economical, durable carboys are resistant to most chemicals. Heavy-duty swing handle allows easy carrying for your liquids and powders. Narrow-Mouth Carboys are complete with rubber O-ring for added security. HDPE closure contains tag holes for identification.
£37.26 - £185.01GBP / Each
£44.71 - £222.01GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material
Availability Pricing
COMPARE
5 High Density Polyethyene (HDPE)
£37.26GBP / Each
£44.71GBP (incl VAT)
COMPARE
10 High Density Polyethyene (HDPE)
£56.75GBP / Each
£68.10GBP (incl VAT)
COMPARE
25 High Density Polyethyene (HDPE)
£91.89GBP / Each
£110.27GBP (incl VAT)
COMPARE
50 High Density Polyethyene (HDPE)
£185.01GBP / Each
£222.01GBP (incl VAT)