Cole-Parmer® Glass Short Stem Funnels
Economical short stemmed glass funnels
  • Borosilicate 3.3 glass
  • Withstands wide temperature range -50 to 550ºC
  • Autoclavable
These glass funnels are ideal for tranferring powders or liquids,
£12.70 - £66.29GBP / Pkg of 6
£15.24 - £79.55GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Stem Length (mm) Stem OD (mm) Top OD (mm)
Availability Pricing
COMPARE
40 7 40
£23.90GBP / Pkg of 16
£28.68GBP (incl VAT)
COMPARE
50 7 50
£17.59GBP / Pkg of 12
£21.11GBP (incl VAT)
COMPARE
60 7 60
£14.36GBP / Pkg of 8
£17.23GBP (incl VAT)
COMPARE
75 9 75
£12.70GBP / Pkg of 6
£15.24GBP (incl VAT)
COMPARE
90 11 90
£31.12GBP / Pkg of 6
£37.34GBP (incl VAT)
COMPARE
100 13 100
£16.85GBP / Pkg of 6
£20.22GBP (incl VAT)
COMPARE
120 15 120
£66.29GBP / Pkg of 8
£79.55GBP (incl VAT)
COMPARE
150 17 150
£21.99GBP / Pkg of 2
£26.39GBP (incl VAT)
Showing 1- 8 of 8