Cole-Parmer® Glass Media Bottles, Class A, Round
Class A round glass media storage bottles ideal for mixing, sampling, and storing mixtures, reagents, solvents, and solutions
  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
Made of borosilicate 3.3 glass, these Class A, round media bottles provide outstanding corrosion resistance and meet ASTM E438 requirements. Able to withstand temperatures from -50 to 550 °C (-58 to 1022 °F), they have a wide mouth opening that allows for easy pouring. The clear, colorless walls have a marking spot for labeling and graduations showing approximate volume. Both the bottle and polypropylene cap are autoclavable.
£17.84 - £203.68GBP / Each
£21.41 - £244.42GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Overall Height (cm)
Availability Pricing
COMPARE
100 20 10.1
£76.58GBP / Case of 10
£91.90GBP (incl VAT)
COMPARE
250 50 13.7
£104.12GBP / Case of 10
£124.94GBP (incl VAT)
COMPARE
500 100 17.5
£140.42GBP / Case of 10
£168.50GBP (incl VAT)
COMPARE
1000 100 22.5
£203.68GBP / Case of 10
£244.42GBP (incl VAT)
COMPARE
2000 200 26
£45.30GBP / Each
£54.36GBP (incl VAT)
COMPARE
25 5 7.5
£17.84GBP / Pkg of 10
£21.41GBP (incl VAT)
COMPARE
50 10 9
£28.54GBP / Pkg of 10
£34.25GBP (incl VAT)
COMPARE
5000 500 33
£79.82GBP / Each
£95.78GBP (incl VAT)
COMPARE
10000 1000 42
£131.50GBP / Each
£157.80GBP (incl VAT)
Showing 1- 9 of 9