Cole-Parmer® Fluorinated HDPE Narrow-Mouth Plastic Bottles
Reduces solvent adsorption and permeation
  • Fluorination enhances long-term container performance
  • Yields lower extractables
These fluorinated narrow-mouth bottles are fluorinated inside and out for improved barrier properties and reduced solvent adsorption and permeation. Bottles include leakproof fluorinated polypropylene (FLPP) screw closures. Split- and puncture-resistant.
£45.39 - £60.98GBP / Pkg of 12
£54.47 - £73.18GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
250 24 Fluorinated High Density Polyethylene (FLPE)
£45.39GBP / Pkg of 12
£54.47GBP (incl VAT)
COMPARE
500 28 Fluorinated High Density Polyethylene (FLPE)
£60.57GBP / Pkg of 12
£72.68GBP (incl VAT)
COMPARE
1000 38-430 Fluorinated High Density Polyethylene (FLPE)
£60.98GBP / Pkg of 6
£73.18GBP (incl VAT)