Cole-Parmer® Economy Wide-Mouth Plastic Bottles, PPCO
Save money while keeping samples contained
  • Lightweight design minimizes added weight
  • Leakproof PP closures
  • Autoclavable
These lightweight bottles are suitable for most laboratory applications. Ideal for light-duty general purpose applications where heavier bottles are not required.
£16.93 - £184.18GBP / Pkg of 12
£20.32 - £221.02GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28 mm Polypropylene Copolymer
£19.17GBP / Pkg of 12
£23.00GBP (incl VAT)
COMPARE
30 28 mm Polypropylene Copolymer
£113.04GBP / Pkg of 72
£135.65GBP (incl VAT)
COMPARE
60 28 mm Polypropylene Copolymer
£16.93GBP / Pkg of 12
£20.32GBP (incl VAT)
COMPARE
60 28 mm Polypropylene Copolymer
£95.20GBP / Pkg of 72
£114.24GBP (incl VAT)
COMPARE
125 43 mm Polypropylene Copolymer
£23.59GBP / Pkg of 12
£28.31GBP (incl VAT)
COMPARE
250 43 mm Polypropylene Copolymer
£35.60GBP / Pkg of 12
£42.72GBP (incl VAT)
COMPARE
250 43 mm Polypropylene Copolymer
£184.18GBP / Pkg of 72
£221.02GBP (incl VAT)
COMPARE
500 53 mm Polypropylene Copolymer
£174.43GBP / Pkg of 48
£209.32GBP (incl VAT)
COMPARE
1000 63 mm Polypropylene Copolymer
£40.57GBP / Pkg of 6
£48.68GBP (incl VAT)
COMPARE
1000 63 mm Polypropylene Copolymer
£169.04GBP / Pkg of 24
£202.85GBP (incl VAT)
Showing 1- 10 of 10