Cole-Parmer® Economy Pre-Cleaned Narrow-Mouth Bottles, Amber Glass
Save time—no need to prewash bottles
  • Pre-cleaned by vacuum and ionization
  • Individually sealed for long term storage with no contamination
The vacuum and ionization process removes contaminants such as dirt, dust, carton lint, oil, aerosols, and fine glass particles from these bottles. Ideal for chemical and resin sampling. Bottles come with PTFE lined caps.
£82.88 - £108.89GBP / Case of 24
£99.46 - £130.67GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 22 mm Glass
£93.34GBP / Case of 24
£112.01GBP (incl VAT)
COMPARE
250 24 mm Glass
£108.89GBP / Case of 24
£130.67GBP (incl VAT)
COMPARE
500 28 mm Glass
£82.88GBP / Case of 12
£99.46GBP (incl VAT)
COMPARE
1000 33 mm Glass
£97.69GBP / Case of 12
£117.23GBP (incl VAT)