Cole-Parmer® Economy Narrow-Mouth Plastic Bottles, PPCO
Store chemical samples safely
  • Leakproof, polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Ideal for storing chemicals and distilled water. Larger bottles feature built-in shoulder loop.
£16.10 - £41.82GBP / Pkg of 12
£19.32 - £50.18GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
60 20 Polypropylene Copolymer
£16.10GBP / Pkg of 12
£19.32GBP (incl VAT)
COMPARE
125 24 Polypropylene Copolymer
£22.74GBP / Pkg of 12
£27.29GBP (incl VAT)
COMPARE
250 24 Polypropylene Copolymer
£31.70GBP / Pkg of 12
£38.04GBP (incl VAT)
COMPARE
500 28 Polypropylene Copolymer
£25.39GBP / Pkg of 6
£30.47GBP (incl VAT)
COMPARE
1000 38-430 Polypropylene Copolymer
£41.82GBP / Pkg of 6
£50.18GBP (incl VAT)