Cole-Parmer® Durable Tube Racks, POM
Highly resistant to acids, bases, solvents, and heat
  • Stable design ideal for use in water baths
  • Flat labeling area makes for easy identification
  • Molded coordinates for tube orientation
  • Chemical and heat resistance
  • Withstands temperatures down to -40 °C
  • Autoclavable at 121 °C
These durable tube racks are molded in a single, continuous piece of fiberglass-reinforced polyoxymethylene (POM). There are no detachable pieces and no assembly required. Racks are chemical and heat resistance with a temperature range of -40 to 121 °C (-40 to 249.8 °F). Teardrop-shaped openings provide better retention of test tubes.
Available in half and full rack sizes for five different tube diameters and three-color options.
£51.03 - £169.46GBP / Case of 8
£61.24 - £203.35GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mL) Tubes Held
Availability Pricing
COMPARE
White 10 36
£51.03GBP / Case of 8
£61.24GBP (incl VAT)
COMPARE
Blue 10 36
£51.03GBP / Case of 8
£61.24GBP (incl VAT)
COMPARE
Green 10 36
£51.03GBP / Case of 8
£61.24GBP (incl VAT)
COMPARE
White 15 36
£110.35GBP / Case of 8
£132.42GBP (incl VAT)
COMPARE
Blue 15 36
£110.35GBP / Case of 8
£132.42GBP (incl VAT)
COMPARE
Green 15 36
£110.35GBP / Case of 8
£132.42GBP (incl VAT)
COMPARE
White 50 9
£119.47GBP / Case of 8
£143.36GBP (incl VAT)
COMPARE
Blue 50 9
£119.47GBP / Case of 8
£143.36GBP (incl VAT)
COMPARE
Green 50 9
£119.47GBP / Case of 8
£143.36GBP (incl VAT)
COMPARE
White 10 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
Blue 10 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
Green 10 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
White 15 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
Blue 15 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
Green 15 72
£153.90GBP / Case of 8
£184.68GBP (incl VAT)
COMPARE
White 50 24
£169.46GBP / Case of 8
£203.35GBP (incl VAT)
COMPARE
Blue 50 24
£169.46GBP / Case of 8
£203.35GBP (incl VAT)
COMPARE
Green 50 24
£169.46GBP / Case of 8
£203.35GBP (incl VAT)
Showing 1- 18 of 18