Cole-Parmer® Digital Bath
Pair with the Stuart rotary evaporators for maximum efficiency and smallest footprint
  • Precise digital temperature display
  • Corrosion-resistant bowl
  • Easy to clean
  • Integrated carry handle for water bath
  • Cool-touch outer case
The clear LED digital display shows the actual bath temperature or the set temperature. There are also two water bath options. The 90°C bath has a 304 stainless steel bowl and is for water only, suitable for low boiling point applications. The higher temperature 180°C bath has an aluminium bowl with PTFE coating and can be used with both water and oil, ideal for higher boiling point liquids.
Note: Use only water bath model 16340-64 with plastic-coated glassware since it is limited to 80°C.
£994.48GBP / Each
£1,193.38GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Description
Availability Pricing
COMPARE
230 4.3 Stuart® Digital Water Bath, 4.3 L, 230 V, 50 Hz
Call For Price
COMPARE
230 5 Stuart® Digital High Temperature Oil Bath, 5 L, 230 V, 50-60 Hz
£994.48GBP / Each
£1,193.38GBP (incl VAT)