Cole-Parmer® Cryogenic Vial Workstations
Effortlessly open vials with one hand
  • Constructed of autoclavable polypropylene
  • Suitable for a temperature range of -196°C to +121°C
Securely holds up to 40 self-standing cryogenic vials. Unique locking design of wells allows for one-handed opening and closing of caps.
£32.52 - £33.31GBP / Each
£39.02 - £39.97GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubes Held Description
Availability Pricing
COMPARE
Blue 40 PolarSafe Cryovial Rack, 40, Blue; 1/Ea
£33.31GBP / Each
£39.97GBP (incl VAT)
COMPARE
Red 40 PolarSafe Cryovial Rack, 40, Red; 1/Ea
£32.52GBP / Each
£39.02GBP (incl VAT)