Cole-Parmer® Cleanroom Stationary Male Luer Lock to Barb Fittings, Straight Adapter, CrystalVu™
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Connect to a female luer body to form a leak-free fluid path
  • Single barb design tightly “grips” the tubing
  • USP Class VI, EU 2002/72/EC, and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. A transparent alternative to polycarbonate, CrystalVu™ fittings do not contain any bisphenol-A (BPA) that has been found to be harmful to humans. These fittings have excellent chemical resistance, withstand temperatures up to 180°C (356°F), and can be autoclaved multiple times with no crazing.
£4.60 - £6.09GBP / Pkg of 10
£5.52 - £7.31GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Luer Connection Type (Port 1)
Availability Pricing
COMPARE
3/32 Polycarbonate Male Luer
£4.62GBP / Pkg of 10
£5.54GBP (incl VAT)
COMPARE
1/16 Polycarbonate Male Luer
£4.62GBP / Pkg of 10
£5.54GBP (incl VAT)
COMPARE
5/32 Polycarbonate Male Luer
£4.94GBP / Pkg of 10
£5.93GBP (incl VAT)
COMPARE
1/8 Polycarbonate Male Luer
£4.60GBP / Pkg of 10
£5.52GBP (incl VAT)
COMPARE
3/16 Polycarbonate Male Luer
£5.15GBP / Pkg of 10
£6.18GBP (incl VAT)
COMPARE
1/4 Polycarbonate Male Luer
£6.09GBP / Pkg of 10
£7.31GBP (incl VAT)