Cole-Parmer® Cleanroom Port Disk Hose Barb Fittings, Flat Plate, Polypropylene
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Flexible, smooth flat base for easy attachment
  • Ultrasonic, radio frequency, or thermal weld to polypropylene bags
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free port disks with hose barb to transfer fluids, buffers, media, and drug compounds from single-use bags used in pharmaceutical and medical industries. Ultrasonic, radio frequency, or thermal weld port disks to polypropylene medical and bioreactor bags to easily transfer fluids.
Animal-derivative-free polypropylene is radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Disks are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
£20.45 - £26.67GBP / Pkg of 10
£24.54 - £32.00GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Color Fitting Material
Availability Pricing
COMPARE
5/8 Natural Polypropylene
£22.52GBP / Pkg of 10
£27.02GBP (incl VAT)
COMPARE
1 Natural Polypropylene
£26.67GBP / Pkg of 10
£32.00GBP (incl VAT)
COMPARE
1/4 Natural Polypropylene
£20.45GBP / Pkg of 10
£24.54GBP (incl VAT)
COMPARE
3/8 Natural Polypropylene
£21.78GBP / Pkg of 10
£26.14GBP (incl VAT)
COMPARE
1/2 Natural Polypropylene
£21.78GBP / Pkg of 10
£26.14GBP (incl VAT)