Cole-Parmer® Cleanroom Male Luer Slip Plugs, Straight Adapter
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Seal luer ports with a secure and leak-proof connection
  • Ideal for capping off fluid transfer system
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
£5.41 - £10.38GBP / Pkg of 10
£6.49 - £12.46GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
COMPARE
Polycarbonate Male Luer Slip Plug
£6.82GBP / Pkg of 10
£8.18GBP (incl VAT)
COMPARE
Nylon Male Luer Slip Plug
£8.52GBP / Pkg of 10
£10.22GBP (incl VAT)
COMPARE
PVDF Male Luer Slip Plug
£10.38GBP / Pkg of 10
£12.46GBP (incl VAT)
COMPARE
Polypropylene Male Luer Slip Plug
£5.41GBP / Pkg of 10
£6.49GBP (incl VAT)
COMPARE
Nylon Male Luer Slip Plug
£5.86GBP / Pkg of 10
£7.03GBP (incl VAT)