Cole-Parmer® 81-Place Polypropylene Storage Boxes
Reinforced hinges, lock, and seal for secure storage
  • Bold, black alpha-numeric printing allows accurate tracking of samples
  • Click fit lock ensures box will not open accidentally—even if placed upside-down
  • Large clasp allows easy opening with gloves
  • Fit in most 2" freezer racks
Boxes withstand temperatures from -80°C to 121°C, and will not degrade with repeated thaws. Each box features an alphanumeric grid on the exterior and interior, stacking rings, and a matte surface suitable for markers or labels. Box top is clear for easy visual inspection of samples and bottoms are colored for additional identification and organization. Boxes accommodate 0.5, 1.5, and 2.0 mL microtubes, and most brands of cryogenic vials.
£47.67 - £435.12GBP / Pkg of 5
£57.20 - £522.14GBP (incl VAT)
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Description
Availability Pricing
COMPARE
Natural (clear) 81-Place Freezer Boxes, Natural, PP; 5/Pk
£49.70GBP / Pkg of 5
£59.64GBP (incl VAT)
COMPARE
Blue 81-Place Freezer Boxes, Blue, PP; 5/Pk
£47.67GBP / Pkg of 5
£57.20GBP (incl VAT)
COMPARE
Green 81-Place Freezer Boxes, Green;, PP; 5/Pk
£49.70GBP / Pkg of 5
£59.64GBP (incl VAT)
COMPARE
Purple 81-Place Freezer Boxes, Purple, PP; 5/Pk
£49.70GBP / Pkg of 5
£59.64GBP (incl VAT)
COMPARE
Orange 81-Place Freezer Boxes, Orange, PP; 5/Pk
Call For Price
COMPARE
Assorted (one of each): natural, blue, green, purple, and orange 81-Place Freezer Boxes, Assorted Colors, PP; 5/Pk
£49.62GBP / Pkg of 5
£59.54GBP (incl VAT)
COMPARE
Green 81-Place Freezer Boxes, Green;, PP; 50/CS
£426.97GBP / Case of 50
£512.36GBP (incl VAT)
COMPARE
Natural (clear) 81-Place Freezer Boxes, Natural, PP; 50/CS
£435.12GBP / Case of 50
£522.14GBP (incl VAT)
COMPARE
Assorted (one of each): natural, blue, green, purple, and orange 81-Place Freezer Boxes, Assorted Colors, PP; 50/CS
£426.97GBP / Case of 50
£512.36GBP (incl VAT)
COMPARE
Blue 81-Place Freezer Boxes, Blue, PP; 50/CS
£426.97GBP / Case of 50
£512.36GBP (incl VAT)
COMPARE
Purple 81-Place Freezer Boxes, Purple, PP; 50/CS
£426.97GBP / Case of 50
£512.36GBP (incl VAT)
COMPARE
Orange 81-Place Freezer Boxes, Orange, PP; 50/CS
£426.97GBP / Case of 50
£512.36GBP (incl VAT)
Showing 1- 12 of 12