Cole-Parmer® 4-Way Stopcocks with Luer Fittings, Gamma-Stable Polycarbonate
Select from four fluid path configurations with a simple turn of the handle
  • Accurate, leak-free flow control
  • Handles rotate a full 360°
  • Transparent polycarbonate lets you easily view fluid flow
  • Conforms to ISO 594-1 and 594-2 standards
Four-way stopcocks feature 3 ports with a 360° rotating handle. Simply adjust the handle to be parallel with two or more ports to select one of four flow configurations. Stopcocks feature Luer fittings, making it easy to create a quick, leak-free connection. Manufactured from high-performance thermoplastic, DEHP-free and RoHS-compliant gamma-stable polycarbonate stopcocks are ideal for applications requiring biocompatibility and stability to gamma irradiation.
£13.04 - £17.54GBP / Pkg of 10
£15.65 - £21.05GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Luer Connection Type (Port 1) Luer Connection Type (Port 2) Luer Connection Type (Port 3)
Availability Pricing
COMPARE
Female Luer Female Luer Female Luer
£13.04GBP / Pkg of 10
£15.65GBP (incl VAT)
COMPARE
Female Luer Male Locking Luer Female Luer
£17.54GBP / Pkg of 10
£21.05GBP (incl VAT)
COMPARE
Female Luer Male Slip Luer Female Luer
£13.04GBP / Pkg of 10
£15.65GBP (incl VAT)
COMPARE
Male Locking Luer Female Luer Female Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Female Luer Male Locking Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Female Luer Male Slip Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Male Locking Luer Female Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Male Locking Luer Male Locking Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Male Slip Luer Female Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Male Slip Luer Male Locking Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Locking Luer Male Slip Luer Male Slip Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Slip Luer Female Luer Female Luer
£13.04GBP / Pkg of 10
£15.65GBP (incl VAT)
COMPARE
Male Slip Luer Female Luer Male Slip Luer
£13.04GBP / Pkg of 10
£15.65GBP (incl VAT)
COMPARE
Male Slip Luer Male Locking Luer Male Locking Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Slip Luer Male Locking Luer Male Slip Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Slip Luer Male Locking Luer Female Luer
£15.86GBP / Pkg of 10
£19.03GBP (incl VAT)
COMPARE
Male Slip Luer Male Slip Luer Female Luer
£13.04GBP / Pkg of 10
£15.65GBP (incl VAT)
COMPARE
Male Slip Luer Male Slip Luer Male Slip Luer
£14.48GBP / Pkg of 10
£17.38GBP (incl VAT)
Showing 1- 18 of 18