Cole-Parmer® 2D Polycarbonate Cryo-Boxes
Make sample processing even easier with automatic identification
  • Constructed of durable polycarbonate for cryogenic applications
  • Ideal for fast and accurate sample processing
The polycarbonate storage boxes work in conjunction with the 2D tubes to provide automatic identification of both the box and stored vials. Each box features a 2D Datamatrix code on the bottom of the box as well as a laser engraved Code 128 barcode on the side. These boxes are available in "short" (for 2mL tubes) or "tall" designs (for 5mL tubes). White grid and blue grid options are available for quick and easy identification.
£28.60 - £30.48GBP / Each
£34.32 - £36.58GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubes Held Tube Size
Availability Pricing
COMPARE
Blue Grid 81 2mL
£28.60GBP / Each
£34.32GBP (incl VAT)
COMPARE
White Grid 100 2mL
£30.07GBP / Each
£36.08GBP (incl VAT)
COMPARE
White Grid 100 5mL
£30.48GBP / Each
£36.58GBP (incl VAT)