BrandTech® BIO-CERT® PCR Tubes with Caps
Reduce evaporation losses with tightly closing cap strips
  • BIO-CERT® PCR QUALITY—Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
  • Thin uniform wall thickness for precise thermal transfer
  • Tightly closing caps protect samples against evaporation
  • Colored tubes available for easy identification of samples
BRAND® individual PCR tubes and 8-strip PCR tubes feature extra thin-walls to ensure optimum heat transfer during PCR, for shorter cycle times over a wide range of temperatures. To prevent sample losses from heating, caps reliably seal the vessels with their tight fit. This ensures quality and repeatable results even with small sample volumes. At the same time, the caps can be opened and closed with ease. Domed caps allow for extra sealing pressure via the lid of the thermal cycler and flat caps are required for qPCR applications. Attached cap strips can be easily opened and closed with one hand, while separate caps are especially suitable for automation, as they won't obstruct robotics.
No matter which cap you choose, all variants seal the vessels reliably, effectively protecting your samples from evaporation and contamination. Tubes are available in several colors, allowing easy sample tracking at a glance.
£77.96 - £319.57GBP / Pkg of 1000
£93.55 - £383.48GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Plate Color Well Volume (mL)
Availability Pricing
BrandTech BIO-CERT® PCR Tube with Flat Cap, Polypropylene, Polypropylene, Pink, 0.2 mL; 1000/Pk 0790535 Mfr # 781301
COMPARE
Individual PCR tubes with attached flat cap Pink 0.2
Call For Price
BrandTech BIO-CERT® PCR Tube with Flat Cap, Polypropylene, Blue, 0.2 mL; 1000/Pk 0790539 Mfr # 781304
COMPARE
Individual PCR tubes with attached flat cap Blue 0.2
Call For Price
BrandTech BIO-CERT® PCR Tube with Flat Cap, Polypropylene, Clear, 0.2 mL; 1000/Pk 0790540 Mfr # 781305
COMPARE
Individual PCR tubes with attached flat cap Clear 0.2
£77.96GBP / Pkg of 1000
£93.55GBP (incl VAT)
BrandTech BIO-CERT® PCR Tube with Flat Cap, Polypropylene, Clear, 0.5 mL; 1000/Pk 0790541 Mfr # 781310
COMPARE
Individual PCR tubes with attached flat cap Clear 0.5
Call For Price
BrandTech BIO-CERT® PCR Tube with Flat Cap, Polypropylene, Yellow, 0.5 mL; 1000/Pk 0790543 Mfr # 781312
COMPARE
Individual PCR tubes with attached flat cap Yellow 0.5
Call For Price
BrandTech BIO-CERT® PCR 8-Strip Tube with Flat Cap, Polypropylene, Clear, 0.2 mL; 250/Pk 0790555 Mfr # 781326
COMPARE
8-strip PCR tubes with detached flat cap strip Clear 0.2
£319.57GBP / Pkg of 250
£383.48GBP (incl VAT)
BrandTech BIO-CERT® PCR 8-Strip Tube with Domed Cap, Polypropylene, Clear, 0.2 mL; 250/Pk 0790557 Mfr # 781327
COMPARE
8-strip PCR tubes with detached domed cap strip Clear 0.2
£300.07GBP / Pkg of 250
£360.08GBP (incl VAT)
Showing 1- 7 of 7