BrandTech® BIO-CERT® PCR Plates
High quality polymerase chain reaction plastics free from leachables
  • Standard 0.2 mL and low-profile 0.15 mL plates and strips Individual tubes available in 0.5 and 0.2 mL sizes
  • Autoclavable, and RNase-, DNase, and DNA-free
  • Thin uniform wall thickness for precise thermal transfer
  • White polymerase chain reaction or PCR models feature smooth surfaces to optimize reflection of fluorescent signals for quantitative Real Time PCR (qPCR)
  • Plates have rigid decks, unique blue alphanumeric coding and cut corner markings for easy identification
  • Cap tool for reliable, soft touch sealing and detaching PCR caps
These multiwell microplates, individual tubes and tube strips, along with strip caps are made of top-quality virgin resins without lubricants or chemical additives. They are an excellent choice for producing reproducible results in nearly all PCR applications. The clear or white plates come in 24-, 48-, and 96-standard and low-profile wells, and 384-well configurations.
White PCR plates and strips offer better results in the analysis of fluorescence signals and quantification of the replicated DNA.
£90.53 - £486.29GBP / Pkg of 50
£108.64 - £583.55GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Plate Color Plate Type
Availability Pricing
BrandTech BIO-CERT® PCR Plate, 384-Well, Clear, Full Skirt, Low-Profile, Non-Elevated Well Rim, 0.03 mL; 50/Pk 0790569 Mfr # 781345
COMPARE
384 Clear Low profile; Full-skirted
£462.00GBP / Pkg of 50
£554.40GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, No Skirt, Standard Profile, Elevated Well Rim, 0.2 mL; 50/Pk 0790571 Mfr # 781350
COMPARE
96 Clear Standard; Without skirt; Elevated well rim
£253.67GBP / Pkg of 50
£304.40GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 384-Well, White, Full Skirt, Low-Profile, Non-Elevated Well Rim, 0.03 mL; 50/Pk 0790574 Mfr # 781358
COMPARE
384 White Low profile; Full-skirted
£486.29GBP / Pkg of 50
£583.55GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, No Skirt, Low-Profile, Non-Elevated Well Rim, 0.15 mL; 50/Pk 0790575 Mfr # 781366
COMPARE
96 Clear Low profile; Without skirt
£238.98GBP / Pkg of 50
£286.78GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, White, No Skirt, Low-Profile, Non-Elevated Well Rim, 0.15 mL; 50/Pk 0790576 Mfr # 781367
COMPARE
96 White Low profile; Without skirt
£271.93GBP / Pkg of 50
£326.32GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, No Skirt, Standard Profile, Non-Elevated Well Rim, 0.2 mL; 50/Pk 0790577 Mfr # 781368
COMPARE
96 Clear Standard; Without skirt
£238.98GBP / Pkg of 50
£286.78GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, Semi-Skirted, Low-Profile, Non-Elevated Well Rim, 0.15 mL; 50/Pk 0790579 Mfr # 781371
COMPARE
96 Clear Low profile; Semi-skirted
£238.98GBP / Pkg of 50
£286.78GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, White, Semi-Skirted, Low-Profile, Non-Elevated Well Rim, 0.15 mL; 50/Pk 0790580 Mfr # 781372
COMPARE
96 White Low profile; Semi-skirted
£271.93GBP / Pkg of 50
£326.32GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, Semi-Skirted, Standard Profile, Non-Elevated Well Rim, 0.2 mL; 50/Pk 0790583 Mfr # 781375
COMPARE
96 Clear Standard; Semi-skirted
£238.98GBP / Pkg of 50
£286.78GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, White, Semi-Skirted, Standard Profile, Non-Elevated Well Rim, 0.2 mL; 50/Pk 0790584 Mfr # 781376
COMPARE
96 White Standard; Semi-skirted
£271.93GBP / Pkg of 50
£326.32GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, Clear, Full Skirt, Low-Profile, Non-Elevated Rim, 0.15 mL; 50/Pk 0790585 Mfr # 781377
COMPARE
96 Clear Low profile; Full-skirted
£404.63GBP / Pkg of 50
£485.56GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 96-Well, White, Full Skirt, Low-Profile, Non-Elevated Well Rim, 0.15 mL; 50/Pk 0790586 Mfr # 781378
COMPARE
96 White Low profile; Full-skirted
£412.97GBP / Pkg of 50
£495.56GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 24-Well, Clear, No Skirt, Standard Profile, Elevated Well Rim, 0.2 mL; 40/Pk 0790589 Mfr # 781411
COMPARE
24 Clear Standard; Without skirt; Elevated well rim
£92.84GBP / Pkg of 40
£111.41GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 48-Well, Clear, No Skirt, Standard Profile, Elevated Well Rim, 0.2 mL; 20/Pk 0790593 Mfr # 781415
COMPARE
48 Clear Standard; Without skirt; Elevated well rim
£90.53GBP / Pkg of 20
£108.64GBP (incl VAT)
BrandTech BIO-CERT® PCR Plate, 48-Well, White, No Skirt, Standard Profile, Elevated Well Rim, 0.2 mL; 20/Pk 0790594 Mfr # 781416
COMPARE
48 White Standard; Without skirt; Elevated well rim
£104.05GBP / Pkg of 20
£124.86GBP (incl VAT)
Showing 1- 15 of 15