BrandTech® BIO-CERT® PCR 8-Strip Caps
Effective protection against evaporation and contamination
  • Easy to open and close without risk of contamination
  • The right cap shape for your application
  • BIO-CERT® PCR QUALITY—Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
With PCR cap strips you can seal PCR strips and plates with 24, 48, and 96 wells reliably and protect your samples effectively against evaporation and contamination.
The shape of the cap strips is optimally matched to BRAND® PCR tube strips and plates so they are easy to open and close. To prevent contamination, the cap strips are equipped with two side tabs for exact positioning before closing and a small opening attachment on each cap. This prevents contact with the inside of the cap. The highly transparent flat cap strips are ideal for qPCR because the cap shape allows accurate fluorescence measurement. With domed cap strips, thermal cyclers without pressure sensors exert a stronger force, thus further increasing the sealing effect.
Because they are manufactured under cleanroom conditions, cap strips are free of DNase, DNA, RNase, and endotoxins.
£42.07 - £52.77GBP / Pkg of 125
£50.48 - £63.32GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Lid (Included) Plate Color Description
Availability Pricing
BrandTech BIO-CERT® PCR 8-Strip Caps, Polypropylene, Flat Cap, Clear; 125/Pk 0790559 Mfr # 781334
COMPARE
Flat cap Clear PCR 8-Strip Caps, Polypropylene, Flat Cap, Clear; 125/Pk
£52.77GBP / Pkg of 125
£63.32GBP (incl VAT)
BrandTech BIO-CERT® PCR 8-Strip Caps, Polypropylene, Domed Cap, Clear; 125/Pk 0790561 Mfr # 781340
COMPARE
Domed cap Clear PCR 8-Strip Caps, Polypropylene, Domed Cap, Clear; 125/Pk
£42.07GBP / Pkg of 125
£50.48GBP (incl VAT)
BrandTech BIO-CERT® PCR 8-Strip Caps, Polypropylene, Domed Cap, Blue; 125/Pk 0790567 Mfr # 781344
COMPARE
Domed cap Blue PCR 8-Strip Caps, Polypropylene, Domed Cap, Blue; 125/Pk
£42.07GBP / Pkg of 125
£50.48GBP (incl VAT)
BrandTech BIO-CERT® PCR 8-Strip Caps for Plates, Polypropylene, Flat Cap, Clear; 300/Pk 0790591 Mfr # 781413
COMPARE
Flat cap for plates Clear PCR 8-Strip Caps for Plates, Polypropylene, Flat Cap, Clear; 300/Pk