Boekel Digital Mechanical Convection Incubators
Multifunctional door design lets you choose between a see-through or a solid window
  • Mechanical convection provides excellent temperature stability, uniformity, and recovery time
  • Wide temperature range accommodates multiple applications
  • Bright, easy-to-use digital display with built-in timer
Use these digital mechanical convection incubators with a wide temperature range for various microbiological testing, wastewater and water quality testing, and general sample incubation applications. A large polycarbonate window makes it easy to view your samples during incubation. The unique door design accepts a tight fitting, easily removeable solid door panel (included) that prevents light from entering the incubator—ideal for light sensitive samples.
These digital incubators use a programmable logic controller (PLC) microprocessor digital control system to maintain accurate and stable temperature. Bright digital display with built-in timer makes it simple to set and view the temperature.
£1,688.30 - £3,123.56GBP / Each
£2,025.96 - £3,748.27GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Capacity (Liters) Power (VAC)
Availability Pricing
Boekel Mechanical Convection Incubator, 14 L (0.5 cu ft); 115 VAC 0110006 Mfr # 151050
COMPARE
14 0.5 115
£1,688.30GBP / Each
£2,025.96GBP (incl VAT)
Boekel Mechanical Convection Incubator, 14 L (0.5 cu ft); 230 VAC 0110007 Mfr # 151050-2
COMPARE
14 0.5 230
£1,694.62GBP / Each
£2,033.54GBP (incl VAT)
Boekel Mechanical Convection Incubator, 40 L (1.4 cu ft); 115 VAC 0110008 Mfr # 151140
COMPARE
40 1.4 115
£1,916.45GBP / Each
£2,299.74GBP (incl VAT)
Boekel Mechanical Convection Incubator, 40 L (1.4 cu ft); 230 VAC 0110009 Mfr # 151140-2
COMPARE
40 1.4 230
£2,011.86GBP / Each
£2,414.23GBP (incl VAT)
Boekel Mechanical Convection Incubator, 71 L (2.5 cu ft); 115 VAC 0110011 Mfr # 151250
COMPARE
71 2.5 115
£2,932.75GBP / Each
£3,519.30GBP (incl VAT)
Boekel Mechanical Convection Incubator, 71 L (2.5 cu ft); 230 VAC 0110012 Mfr # 151250-2
COMPARE
71 2.5 230
£2,820.75GBP / Each
£3,384.90GBP (incl VAT)
Boekel Mechanical Convection Incubator, 141 L (5.0 cu ft); 115 VAC 0110013 Mfr # 151500
COMPARE
141 5 115
£3,032.30GBP / Each
£3,638.76GBP (incl VAT)
Boekel Mechanical Convection Incubator, 141 L (5.0 cu ft); 230 VAC 0110014 Mfr # 151500-2
COMPARE
141 5 230
£3,123.56GBP / Each
£3,748.27GBP (incl VAT)
Showing 1- 8 of 8