Alnor Low Flow Balometer Capture Hood
Analog bar graph monitors air trends simultaneously with a digital display
  • Ideal for light commercial ventilation measurements
This digital balometer features a 24” x 24” hood opening to fit over standard-sized grilles and diffusers located on ceilings, walls or floors. The hood channels airflow through the base to directly measure air volume (CFM). Weighing less than six pounds, the balometer allows for single operator use and access into tight spaces.
Fast response LCD features an analog bar graph for monitoring airflow changes and displays digital readout. Measure air volume with or without hood connected to the balometer.
What’s included: NIST-traceable calibration certificate, and four C batteries.
£1,957.93GBP / Each
£2,349.52GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Air Volume (CFM) Volume Accuracy Volume Resolution
Availability Pricing
TSI G200D Low Flow Balometer Capture Hood, 2 ft X 2 ft 1055022 Mfr # 6200D
COMPARE
500 ±3% + (5 CFM or 8.5 m³/h) 1 CFM (1 m³/h, 4 L/s)
£1,957.93GBP / Each
£2,349.52GBP (incl VAT)