Alicat MB-Series Mass Flowmeters with Digital Display, Battery Operated, SLPM
Provides fast, stable, repeatable flow rate control in milliseconds
  • Built-in rechargeable battery with up to 18 hours life
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • IP40 rating
  • Limited lifetime warranty
These portable mass flowmeters, with a fast control response time of 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They come with 98 pre-loaded gas calibrations, which can be switched seamlessly while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure with flow rates displayed in standard liters per minute (SLPM). Key features include a rechargeable battery, backlit monochrome display, data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
£1,162.97 - £4,533.34GBP / Each
£1,395.56 - £5,440.01GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Flow Rate (LPM) Repeatability
Availability Pricing
Alicat Mass Flowmeter with Display, 0 - 1 SLPM, Battery Operated 6780279 Mfr # MB-1SLPM-D
COMPARE
±0.6% of reading 1 ±0.1% of reading + 0.02% of full scale
£1,162.97GBP / Each
£1,395.56GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 2 SLPM, Battery Operated 6780280 Mfr # MB-2SLPM-D
COMPARE
±0.6% of reading 2 ±0.1% of reading + 0.02% of full scale
£1,162.97GBP / Each
£1,395.56GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 5 SLPM, Battery Operated 6780281 Mfr # MB-5SLPM-D
COMPARE
±0.6% of reading 5 ±0.1% of reading + 0.02% of full scale
£1,285.20GBP / Each
£1,542.24GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 10 SLPM, Battery Operated 6780282 Mfr # MB-10SLPM-D
COMPARE
±0.6% of reading 10 ±0.1% of reading + 0.02% of full scale
£1,162.97GBP / Each
£1,395.56GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 20 SLPM, Battery Operated 6780283 Mfr # MB-20SLPM-D
COMPARE
±0.6% of reading 20 ±0.1% of reading + 0.02% of full scale
£1,162.97GBP / Each
£1,395.56GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 50 SLPM, Battery Operated 6780284 Mfr # MB-50SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 50 ±0.2% of reading + 0.02% of full scale
£1,233.34GBP / Each
£1,480.01GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 100 SLPM, Battery Operated 6780285 Mfr # MB-100SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 100 ±0.2% of reading + 0.02% of full scale
£1,233.34GBP / Each
£1,480.01GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 250 SLPM, Battery Operated 6780286 Mfr # MB-250SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 250 ±0.2% of reading + 0.02% of full scale
£1,677.78GBP / Each
£2,013.34GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 500 SLPM, Battery Operated 6780287 Mfr # MB-500SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 500 ±0.2% of reading + 0.02% of full scale
£1,677.78GBP / Each
£2,013.34GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 1000 SLPM, Battery Operated 6780288 Mfr # MB-1000SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 1000 ±0.2% of reading + 0.02% of full scale
£2,111.12GBP / Each
£2,533.34GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 2000 SLPM, Battery Operated 6780289 Mfr # MB-2000SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 2000 ±0.2% of reading + 0.02% of full scale
£2,925.93GBP / Each
£3,511.12GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 3000 SLPM, Battery Operated 6780290 Mfr # MB-3000SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 3000 ±0.2% of reading + 0.02% of full scale
£3,318.52GBP / Each
£3,982.22GBP (incl VAT)
Alicat Mass Flowmeter with Display, 0 - 5000 SLPM, Battery Operated 6780291 Mfr # MB-5000SLPM-D
COMPARE
±0.8% of reading + 0.2% of full scale 5000 ±0.2% of reading + 0.02% of full scale
£4,533.34GBP / Each
£5,440.01GBP (incl VAT)
Showing 1- 13 of 13