Ace Glass Tube Adapter
  • Adapter has a Standard Taper bottom joint and #7 Ace threads on top.
Nylon bushing tightens on top threads to form an O-ring compression seal for use with thermometers and bleed tubes up to 7 mm in diameter. All adapters include nylon bushing and FETFE O-ring.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Ace Glass 5028-24 Tube Adapter, 10/18 Joint 3472100 Mfr # 5028-24
COMPARE
Call For Price
Ace Glass 5028-25 Tube Adapter, 14/10 Joint 3472102 Mfr # 5028-25