ASTM Class 1 - Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as0. 1 mg to 0.01 mg .
Surfaces are polished to a perfect, mirror like finish
... READ MORE
Weights 1mg through 10kg are supplied in a high quality polycarbonate case, all weights include noted documentation of accuracy from the vendor.
Key Features
Surfaces are polished to a perfect, mirror like finish
Masses are less vulnerable to wear and scratches
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
More About this Item
Appropriate for calibrating high-precision analytical balances with readability as low as 0.1 mg to 0.01 mg.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to,Manufacturing QC, Secondary Calibration Labs, State Laboratories, Pharmaceutical QC, R&D, Analytical Testing, Chemical Manufacturing, Utilities.
Specifications
Mass ClassClass 1
Mass Size (mg)200
Mass Size (g)0.2
Mass Size (kg)0.0002
Mass Size (lbs)0.00044
Mass CertificationNIST
Tolerance0.010 mg
Height (in)0.125
Length (in)0.27
Description100 mg Class 1 Analytical Weight with Traceable Cert