Thermo Scientific Nalgene BPC3410 Wide Mouth Level 3 Uncertified Natural Color Bottle, Round, Not Preserved, 250 mL; 24/CS
Representative image only.
Thermo Scientific Nalgene BPC3410 Wide Mouth Level 3 Uncertified Natural Color Bottle, Round, Not Preserved, 250 mL; 24/CS
Thermo Scientific | Item: 3521055
For general lab use
£50.86GBP / Case of 24
£61.03GBP (incl VAT)
£50.86GBP / Case of 24
£61.03GBP (incl VAT)
Product Description
Key Features
  • Multiple bottle sizes available
  • Bottles are not preserved
More About this Item
Level 3 bottles are not certified to EPA guidelines for contaminant free containers and do not come with a certificate of analysis.
Specifications
  • Capacity (mL)250
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Level Number3
  • Special FeaturesNot preserved
  • Neck Inside Diameter (ID) (mm)43
  • DescriptionLevel 3 Uncertified Natural Color Bottle, Round, Not Preserved, 250 mL, 24/Case