Thermo Scientific Nalgene BPC3400 Wide Mouth Level 3 Uncertified Natural Color Bottle, Round, Not Preserved, 125 mL; 48/CS
Representative image only.
Thermo Scientific Nalgene BPC3400 Wide Mouth Level 3 Uncertified Natural Color Bottle, Round, Not Preserved, 125 mL; 48/CS
Thermo Scientific | Item: 3521054
For general lab use
£67.09GBP / Case of 48
£80.51GBP (incl VAT)
£67.09GBP / Case of 48
£80.51GBP (incl VAT)
Product Description
Key Features
  • Multiple bottle sizes available
  • Bottles are not preserved
More About this Item
Level 3 bottles are not certified to EPA guidelines for contaminant free containers and do not come with a certificate of analysis.
Specifications
  • Capacity (mL)125
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Level Number3
  • Special FeaturesNot preserved
  • Neck Inside Diameter (ID) (mm)38
  • DescriptionLevel 3 Uncertified Natural Color Bottle, Round, Not Preserved, 125 mL, 48/Case