Thermo Scientific AC45ST Standards kit for AQ4500 Turbidimeter
Representative image only.
Thermo Scientific AC45ST Standards kit for AQ4500 Turbidimeter
Thermo Scientific | Mfr: AC45ST| Item: 9954552
Optimize water analysis
£407.68GBP / Each
£489.22GBP (incl VAT)
£407.68GBP / Each
£489.22GBP (incl VAT)
Buy It Together
Current Item
Thermo Scientific AC45ST Standards kit for AQ4500 Turbidimeter
Thermo Scientific AC45ST Standards kit for AQ4500 Turbidimeter
9954552
£407.68GBP / Each
£489.22GBP (incl VAT)
+
Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
0571870
£653.34GBP / Each
£784.01GBP (incl VAT)
=
Total Price 1061.02 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories for Orion meters
More About this Item
Find electrodes, probes, cables, adapters and much more.
Turbidity standards kit contains 0, 1, 10, 100, and 1000 NTU vials.
Specifications
  • Compatible WithThermo Scientific Orion AQ4500 turbidimeter
  • DescriptionStandards kit for AQ4500 Turbidimeter