Thermo Scientific 700001 Fisher Pure Water pH Test Kit
Representative image only.
Thermo Scientific 700001 Fisher Pure Water pH Test Kit
Thermo Scientific | Mfr: 700001| Item: 5535050
Optimize water analysis
£363.26GBP / Each
£435.91GBP (incl VAT)
£363.26GBP / Each
£435.91GBP (incl VAT)
Buy It Together
Current Item
Thermo Scientific 700001 Fisher Pure Water pH Test Kit
Thermo Scientific 700001 Fisher Pure Water pH Test Kit
5535050
£363.26GBP / Each
£435.91GBP (incl VAT)
+
Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
0571870
£653.34GBP / Each
£784.01GBP (incl VAT)
=
Total Price 1016.60 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories for Orion meters
More About this Item
Find electrodes, probes, cables, adapters and much more.
Provide fast and accurate pH results in high purity or low ionic strength water. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water, and process water.
Contains: 4 x 475mL pH 6.97 Buffer A, 2 x 475mL pH 4.10 Buffer B, 2 x 60mL pHISA Adjuster
Specifications
  • Compatible WithRecommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water
  • DescriptionFisher Pure Water pH Test Kit