Thermo Scientific 083005MD Dissolved Oxygen Probe, Polarographic, 1.5-m Cable
Representative image only.
Thermo Scientific 083005MD Dissolved Oxygen Probe, Polarographic, 1.5-m Cable
Thermo Scientific | Mfr: 083005MD| Item: 5310160
Get accurate dissolved oxygen readings, in the lab or in the field
£1,071.47GBP / Each
£1,285.76GBP (incl VAT)
£1,071.47GBP / Each
£1,285.76GBP (incl VAT)
Product Description
Key Features
  • Compatible with Orion™ Star A series meters
  • Complies with SM 4500-O G and ASTM D888-09 (B)
  • Features automatic temperature compensation (ATC)
  • Convenient screw-on membrane
  • Provides 90% of final value in 10 seconds
  • 9-pin Mini DIM connection
  • 1 year warranty
More About this Item
Choose the probe and parts that meet your application needs.
The polarographic dissolved oxygen (DO) probe features ATC and only drifts about 0.1%/day to provide accurate measurements. Probe requires minimal sample flow of 10 cm/sec.
1 Year Warranty
Specifications
  • Accessory TypePolarographic DO probe
  • Use WithOrion Star series meters
  • DescriptionDissolved Oxygen Probe, Polarographic, 1.5-m Cable