Taylor 5994 Series Refrigerator / Freezer Analog Dial Thermometer with HACCP Guidelines
Representative image only.
Taylor 5994 Series Refrigerator / Freezer Analog Dial Thermometer with HACCP Guidelines
Taylor | Mfr: 5994| Item: 9000130
Ensure proper temperature of your food
Call For Price
for an additional
£89.40GBP / Each
£107.28GBP (incl VAT)
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Taylor 5994 Series Refrigerator / Freezer Analog Dial Thermometer with HACCP Guidelines
Taylor 5994 Series Refrigerator / Freezer Analog Dial Thermometer with HACCP Guidelines
9000130
Call For Price
+
InnoCal NIST-Traceable Calibration, Bimetal or Digital-Pocket Thermometer
1700300
£89.40GBP / Each
£107.28GBP (incl VAT)
=
Total Price 89.40 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3" dial
  • Temperature range -30°F to 120°F (-34°C to 20°C)
  • Safe temperature zone guide
  • Adhesive backing
  • Stainless steel
  • NSF
  • 1 year warranty
More About this Item
5* Refrigerator/Freezer Dial Thermometer features a built-in hanger for secure placement. Large 3" (5.1 cm) dual-scale dial is easy to read and includes a safe temperature zone guide and HACCP guidelines. Durable stainless steel construction with a red pointer.
1 Year WarrantyNSF international certified
Specifications
  • Min Temperature (° F)-30
  • Min Temperature (° C)-30
  • Max Temperature (° F)70
  • Max Temperature (° C)20
  • Accuracy2%
  • DescriptionRefrigerator / Freezer Analog Dial Thermometer with HACCP Guidelines