Sussman 39203C Replacment Heating Element, 20 kW, 240 VAC

Representative image only.

Sussman 39203C Replacment Heating Element, 20 kW, 240 VAC

Sussman | Mfr: 39203C| Item: 0760331

£772.39GBP / Each
£926.87GBP (incl VAT)

£772.39GBP / Each
£926.87GBP (incl VAT)
Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

1 Year Warranty
Specifications
  • Compatible WithSussman Steam Generator with 20 kW, 240 VAC power
  • Power (VAC)240
  • DescriptionReplacment Heating Element, 20 kW, 240 VAC
  • Warranty1 year
Reviews
Q&A